Search
Search

RAP1GAP| Gene Symbol: RAP1GAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130288.2 | XP_014985774.1 | 961 | 93 | ||
| XM_015130302.2 | XP_014985788.1 | 1059 | 93 | |||
| XM_028838395.1 | XP_028694228.1 | 1141 | 93 | |||
| XM_028838396.1 | XP_028694229.1 | 1141 | 93 | |||
| XM_028838412.1 | XP_028694245.1 | 1140 | 93 | |||
| XM_028838420.1 | XP_028694253.1 | 1141 | 93 | |||
| XM_028838425.1 | XP_028694258.1 | 1140 | 93 | |||
| XM_028838429.1 | XP_028694262.1 | 1140 | 93 | |||
| XM_028838431.1 | XP_028694264.1 | 1140 | 93 | |||
| GenBank mRNA | JU320107.1 | - | 924 | 93 | ||
| JU322201.1 | - | 924 | 93 | |||
| JU335753.1 | - | 924 | 93 | |||
| JV600944.1 | - | 944 | 93 | |||
| JV669156.1 | - | 914 | 93 | |||
Set Membership: |
|