Search
Search

KIFC3| Gene Symbol: KIFC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126260.2 | XP_014981746.1 | 1979 | 61 | ||
| XM_015126261.2 | XP_014981747.1 | 1978 | 61 | |||
| XM_015126265.2 | XP_014981751.1 | 3043 | 61 | |||
| XM_015126271.2 | XP_014981757.1 | 1354 | 61 | |||
| XM_028841198.1 | XP_028697031.1 | 1926 | 61 | |||
| XM_028841199.1 | XP_028697032.1 | 1926 | 61 | |||
| XM_028841200.1 | XP_028697033.1 | 1584 | 61 | |||
| XM_028841202.1 | XP_028697035.1 | 1584 | 61 | |||
| XM_028841203.1 | XP_028697036.1 | 1493 | 61 | |||
| GenBank mRNA | JU322116.1 | - | 1399 | 61 | ||
| JV650870.1 | - | 1123 | 61 | |||
Set Membership: |
|