Search
Search

GLG1| Gene Symbol: GLG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126616.2 | XP_014982102.1 | 825 | 102 | ||
| GenBank mRNA | JU327561.1 | - | 555 | 102 | ||
| JU327562.1 | - | 555 | 102 | |||
| JU484582.1 | - | 548 | 102 | |||
Set Membership: |
|