Search
Search

AIG1| Gene Symbol: AIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260565.1 | NP_001247494.1 | 343 | 92 | ||
| XM_028846704.1 | XP_028702537.1 | 483 | 92 | |||
| XM_028846705.1 | XP_028702538.1 | 541 | 92 | |||
| XM_028846706.1 | XP_028702539.1 | 377 | 92 | |||
| XM_028846707.1 | XP_028702540.1 | 457 | 92 | |||
| GenBank mRNA | JU328131.1 | - | 274 | 92 | ||
| JV047297.1 | - | 343 | 92 | |||
| JV601698.1 | - | 336 | 92 | |||