Search
Search

SRSF5| Gene Symbol: SRSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109903.3 | XP_001109903.1 | 510 | 69 | ||
| XM_001110146.4 | XP_001110146.2 | 498 | 69 | |||
| XM_015143974.2 | XP_014999460.1 | 1324 | 69 | |||
| XM_015143975.2 | XP_014999461.1 | 476 | 69 | |||
| XM_015143976.2 | XP_014999462.1 | 510 | 69 | |||
| XM_015143977.2 | XP_014999463.1 | 316 | 69 | |||
| XM_015143978.2 | XP_014999464.1 | 514 | 69 | |||
| GenBank mRNA | JU335901.1 | - | 465 | 69 | ||
| JU335902.1 | - | 336 | 69 | |||
| JU335903.1 | - | 340 | 69 | |||
| JU476773.1 | - | 483 | 69 | |||
| JU476774.1 | - | 354 | 69 | |||
| JU476775.1 | - | 358 | 69 | |||
Set Membership: |
|