Search
Search

NAA25| Gene Symbol: NAA25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152849.2 | XP_015008335.1 | 2804 | 75 | ||
| XM_015152850.2 | XP_015008336.1 | 1239 | 75 | |||
| XM_015152851.2 | XP_015008337.1 | 1055 | 75 | |||
| GenBank mRNA | JU336470.1 | - | 1155 | 75 | ||
| JU336471.1 | - | 1155 | 75 | |||
| JU471444.1 | - | 1158 | 75 | |||
Set Membership: |
|