Search
Search

ZC4H2| Gene Symbol: ZC4H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261736.1 | NP_001248665.1 | 617 | 72 | ||
| XM_015127581.2 | XP_014983067.1 | 777 | 72 | |||
| GenBank mRNA | JU335463.1 | - | 672 | 72 | ||
| JU476405.1 | - | 617 | 72 | |||
Set Membership: |
|