Search
Search

THSD7A| Gene Symbol: THSD7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134011.2 | XP_014989497.2 | 3935 | 57 | ||
| XM_015134012.2 | XP_014989498.2 | 3935 | 57 | |||
| GenBank mRNA | JU333416.1 | - | 3558 | 57 | ||
| JV709592.1 | - | 3566 | 57 | |||
Set Membership: |
|