Search
Search

SCN1B| Gene Symbol: SCN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258167.1 | NP_001245096.1 | 233 | 59 | ||
| XM_015123520.2 | XP_014979006.1 | 839 | 59 | |||
| XM_028838435.1 | XP_028694268.1 | 569 | 59 | |||
| XM_028838436.1 | XP_028694269.1 | 889 | 59 | |||
| XM_028838437.1 | XP_028694270.1 | 568 | 59 | |||
| GenBank mRNA | JU321213.1 | - | 345 | 59 | ||
| JU473653.1 | - | 233 | 59 | |||
| JV667885.1 | - | 347 | 59 | |||
Set Membership: |
|