Search
Search

RASGRP3| Gene Symbol: RASGRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261700.2 | NP_001248629.1 | 1142 | 76 | ||
| XM_015112055.2 | XP_014967541.2 | 1913 | 76 | |||
| XM_015112056.2 | XP_014967542.2 | 1147 | 76 | |||
| XM_015112057.2 | XP_014967543.2 | 1711 | 76 | |||
| XM_015112058.2 | XP_014967544.2 | 1922 | 76 | |||
| XM_015112059.2 | XP_014967545.1 | 1232 | 76 | |||
| XM_028831529.1 | XP_028687362.1 | 1059 | 76 | |||
| XM_028831530.1 | XP_028687363.1 | 951 | 76 | |||
| GenBank mRNA | JU322161.1 | - | 1142 | 76 | ||
Set Membership: |
|