Search
Search

MOK| Gene Symbol: MOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144514.2 | XP_015000000.2 | 438 | 128 | ||
| XM_028851851.1 | XP_028707684.1 | 344 | 128 | |||
| XM_028851852.1 | XP_028707685.1 | 344 | 128 | |||
| XM_028851853.1 | XP_028707686.1 | 344 | 128 | |||
| XM_028851854.1 | XP_028707687.1 | 373 | 128 | |||
| XM_028851855.1 | XP_028707688.1 | 438 | 128 | |||
| XM_028851860.1 | XP_028707693.1 | 344 | 128 | |||
Set Membership: |
|