Search
Search

RIPOR1| Gene Symbol: RIPOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126402.2 | XP_014981888.1 | 3592 | 101 | ||
| XM_015126404.2 | XP_014981890.1 | 3570 | 101 | |||
| XM_015126405.1 | XP_014981891.1 | 3644 | 101 | |||
| XM_028841362.1 | XP_028697195.1 | 4017 | 101 | |||
| XM_028841364.1 | XP_028697197.1 | 3583 | 101 | |||
Set Membership: |
|