Search
Search

C15H9orf24| Gene Symbol: C15H9orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194086.2 | NP_001181015.1 | 1009 | 68 | ||
| XM_015117370.1 | XP_014972856.1 | 1198 | 68 | |||
| XM_015117371.1 | XP_014972857.1 | 1195 | 68 | |||
| XM_015117373.1 | XP_014972859.1 | 1039 | 68 | |||
| XM_015117374.1 | XP_014972860.1 | 471 | 68 | |||
| XM_015117375.2 | XP_014972861.1 | 656 | 68 | |||
Set Membership: |
|