Search
Search

MIIP| Gene Symbol: MIIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113192.2 | XP_014968678.2 | 601 | 64 | ||
| XM_028839677.1 | XP_028695510.1 | 603 | 64 | |||
| GenBank mRNA | JV720567.1 | - | 548 | 64 | ||
Set Membership: |
|