Search
Search

ATP5IF1| Gene Symbol: ATP5IF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260663.1 | NP_001247592.1 | 192 | 62 | ||
| GenBank mRNA | JU472972.1 | - | 192 | 62 | ||
| JV563352.1 | - | 268 | 62 | |||
| JV633820.1 | - | 263 | 62 | |||
Set Membership: |
|