Search
Search

BTBD2| Gene Symbol: BTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122400.2 | XP_014977886.1 | 893 | 56 | ||
| GenBank mRNA | JU328228.1 | - | 1066 | 56 | ||
| JV621648.1 | - | 1103 | 56 | |||
Set Membership: |
|