Search
Search

FGD1| Gene Symbol: FGD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806251.3 | XP_002806297.2 | 2730 | 62 | ||
| XM_028841995.1 | XP_028697828.1 | 1775 | 62 | |||
| XM_028841996.1 | XP_028697829.1 | 1780 | 62 | |||
| GenBank mRNA | JV460466.1 | - | 1437 | 62 | ||
| JV565080.1 | - | 2045 | 62 | |||
Set Membership: |
|