Search
Search

TNS3| Gene Symbol: TNS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133582.2 | XP_014989068.2 | 1007 | 59 | ||
| XM_015133583.2 | XP_014989069.1 | 1060 | 59 | |||
| XM_015133584.2 | XP_014989070.1 | 1016 | 59 | |||
| XM_015133585.2 | XP_014989071.1 | 1069 | 59 | |||
| XM_015133586.2 | XP_014989072.1 | 1035 | 59 | |||
| XM_015133587.2 | XP_014989073.1 | 1001 | 59 | |||
| XM_028845787.1 | XP_028701620.1 | 1020 | 59 | |||
| XM_028845788.1 | XP_028701621.1 | 812 | 59 | |||
| XM_028845789.1 | XP_028701622.1 | 764 | 59 | |||
| XM_028845790.1 | XP_028701623.1 | 982 | 59 | |||
| XM_028845791.1 | XP_028701624.1 | 982 | 59 | |||
| GenBank mRNA | JU335944.1 | - | 988 | 59 | ||
| JU335945.1 | - | 988 | 59 | |||
| JV047950.1 | - | 934 | 59 | |||
| JV047951.1 | - | 934 | 59 | |||
| JV721262.1 | - | 986 | 59 | |||
Set Membership: |
|