Search
Search

ACTN1| Gene Symbol: ACTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143969.2 | XP_014999455.1 | 2855 | 56 | ||
| XM_015143970.2 | XP_014999456.1 | 2584 | 56 | |||
| GenBank mRNA | JU324899.1 | - | 2542 | 56 | ||
| JU470606.1 | - | 2480 | 56 | |||
| JV048568.1 | - | 2502 | 56 | |||
| JV565465.1 | - | 2570 | 56 | |||
Set Membership: |
|