Search
Search

ARHGEF17| Gene Symbol: ARHGEF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115376.4 | XP_001115376.2 | 4050 | 58 | ||
| XM_028833807.1 | XP_028689640.1 | 4051 | 58 | |||
| GenBank mRNA | JV076756.1 | - | 2988 | 58 | ||
| JV666801.1 | - | 3559 | 58 | |||
Set Membership: |
|