Search
Search

VAT1L| Gene Symbol: VAT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841289.1 | XP_028697122.1 | 393 | 66 | ||
| GenBank mRNA | JU322333.1 | - | 322 | 66 | ||
| JV566529.1 | - | 323 | 66 | |||
Set Membership: |
|