Search
Search

EIF4G1| Gene Symbol: EIF4G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131448.2 | XP_014986934.1 | 2720 | 81 | ||
| XM_015131449.2 | XP_014986935.1 | 2958 | 81 | |||
| XM_015131450.2 | XP_014986936.1 | 2653 | 81 | |||
| XM_015131451.2 | XP_014986937.1 | 2763 | 81 | |||
| XM_015131452.2 | XP_014986938.1 | 2706 | 81 | |||
| XM_015131453.2 | XP_014986939.1 | 2745 | 81 | |||
| XM_015131454.2 | XP_014986940.1 | 2937 | 81 | |||
| XM_015131455.2 | XP_014986941.1 | 2632 | 81 | |||
| XM_015131456.2 | XP_014986942.1 | 2742 | 81 | |||
| XM_015131457.2 | XP_014986943.1 | 2780 | 81 | |||
| XM_015131458.2 | XP_014986944.1 | 2793 | 81 | |||
| XM_015131460.2 | XP_014986946.1 | 2630 | 81 | |||
| XM_015131461.2 | XP_014986947.1 | 2795 | 81 | |||
| XM_015131462.2 | XP_014986948.1 | 2009 | 81 | |||
| XM_028843898.1 | XP_028699731.1 | 2455 | 81 | |||
| XM_028843899.1 | XP_028699732.1 | 2504 | 81 | |||
| GenBank mRNA | JU333288.1 | - | 2629 | 81 | ||
| JU333289.1 | - | 2910 | 81 | |||
| JU333290.1 | - | 2889 | 81 | |||
| JU474604.1 | - | 2771 | 81 | |||
| JV046382.1 | - | 2606 | 81 | |||
| JV046383.1 | - | 2663 | 81 | |||
| JV046384.1 | - | 2798 | 81 | |||
| JV608238.1 | - | 2708 | 81 | |||
Set Membership: |
|