Search
Search

POFUT2| Gene Symbol: POFUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132723.2 | XP_014988209.2 | 561 | 62 | ||
| XM_028845238.1 | XP_028701071.1 | 623 | 62 | |||
| XM_028845239.1 | XP_028701072.1 | 561 | 62 | |||
| GenBank mRNA | JV043519.1 | - | 532 | 62 | ||
| JV646488.1 | - | 466 | 62 | |||
Set Membership: |
|