Search
Search

PARD6A| Gene Symbol: PARD6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266564.2 | NP_001253493.1 | 102 | 106 | ||
| XM_015126410.2 | XP_014981896.2 | 182 | 106 | |||
| GenBank mRNA | JV667407.1 | - | 102 | 106 | ||
| JV716151.1 | - | 143 | 106 | |||
Set Membership: |
|