Search
Search

OTUD5| Gene Symbol: OTUD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127377.2 | XP_014982863.2 | 2303 | 64 | ||
| XM_015127378.2 | XP_014982864.2 | 2303 | 64 | |||
| XM_015127379.2 | XP_014982865.2 | 2303 | 64 | |||
| XM_015127380.2 | XP_014982866.2 | 2303 | 64 | |||
| XM_015127381.2 | XP_014982867.2 | 2287 | 64 | |||
| XM_015127382.2 | XP_014982868.2 | 2287 | 64 | |||
| XM_015127383.2 | XP_014982869.2 | 2287 | 64 | |||
| XM_028842464.1 | XP_028698297.1 | 2300 | 64 | |||
| XM_028842465.1 | XP_028698298.1 | 2284 | 64 | |||
| XM_028842466.1 | XP_028698299.1 | 1182 | 64 | |||
| GenBank mRNA | JU328221.1 | - | 1085 | 64 | ||
| JU328223.1 | - | 1100 | 64 | |||
| JV655557.1 | - | 1346 | 64 | |||
| JV655558.1 | - | 1346 | 64 | |||
Set Membership: |
|