Search
Search

TCTA| Gene Symbol: TCTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260695.1 | NP_001247624.1 | 349 | 60 | ||
| GenBank mRNA | JU336123.1 | - | 406 | 60 | ||
| JU336124.1 | - | 406 | 60 | |||
| JU336125.1 | - | 406 | 60 | |||
| JU476986.1 | - | 349 | 60 | |||
Set Membership: |
|