Search
Search

PEX10| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083901.4 | XP_001083901.2 | 1035 | 98 | ||
| XM_015135799.2 | XP_014991285.1 | 859 | 98 | |||
| XM_015135824.2 | XP_014991310.2 | 1363 | 98 | |||
| XM_015135839.2 | XP_014991325.1 | 855 | 98 | |||
| XM_015135886.2 | XP_014991372.1 | 708 | 98 | |||
| XM_028841040.1 | XP_028696873.1 | 906 | 98 | |||
| GenBank mRNA | JU334980.1 | - | 953 | 98 | ||
| JV584387.1 | - | 974 | 98 | |||
Set Membership: |
|