Search
Search

ATP9B| Gene Symbol: ATP9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087685.4 | XP_001087685.3 | 3147 | 65 | ||
| XM_001087804.4 | XP_001087804.2 | 3147 | 65 | |||
| XM_015122229.2 | XP_014977715.2 | 3066 | 65 | |||
| XM_015122230.2 | XP_014977716.2 | 3036 | 65 | |||
| XM_015122231.2 | XP_014977717.2 | 3147 | 65 | |||
| XM_015122232.2 | XP_014977718.2 | 3000 | 65 | |||
| XM_015122235.2 | XP_014977721.2 | 2633 | 65 | |||
| XM_015122236.2 | XP_014977722.2 | 2404 | 65 | |||
| XM_028837950.1 | XP_028693783.1 | 2955 | 65 | |||
| XM_028837951.1 | XP_028693784.1 | 2919 | 65 | |||
| XM_028837952.1 | XP_028693785.1 | 2276 | 65 | |||
| GenBank mRNA | JU321077.1 | - | 3024 | 65 | ||
| JV046996.1 | - | 3037 | 65 | |||
| JV046997.1 | - | 3037 | 65 | |||
| JV650708.1 | - | 2926 | 65 | |||
Set Membership: |
|