Search
Search

SPHK2| Gene Symbol: SPHK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257487.1 | NP_001244416.1 | 804 | 90 | ||
| XM_015124381.2 | XP_014979867.1 | 1515 | 90 | |||
| XM_015124382.2 | XP_014979868.1 | 1524 | 90 | |||
| XM_015124383.2 | XP_014979869.1 | 747 | 90 | |||
| XM_015124384.2 | XP_014979870.1 | 1039 | 90 | |||
| XM_028838491.1 | XP_028694324.1 | 960 | 90 | |||
| GenBank mRNA | JU335508.1 | - | 804 | 90 | ||
Set Membership: |
|