Search
Search

RGS12| Gene Symbol: RGS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137901.2 | XP_014993387.1 | 3135 | 83 | ||
| XM_015137902.2 | XP_014993388.2 | 1309 | 83 | |||
| XM_015137903.2 | XP_014993389.2 | 1474 | 83 | |||
| XM_028848273.1 | XP_028704106.1 | 3255 | 83 | |||
| GenBank mRNA | JU325620.1 | - | 1685 | 83 | ||
| JU473611.1 | - | 3139 | 83 | |||
Set Membership: |
|