Search
Search

RGS12| Gene Symbol: RGS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137901.2 | XP_014993387.1 | 2094 | 92 | ||
| XM_028848273.1 | XP_028704106.1 | 2214 | 92 | |||
| GenBank mRNA | JU325620.1 | - | 644 | 92 | ||
| JU473611.1 | - | 2098 | 92 | |||