Search
Search

SPIN1| Gene Symbol: SPIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118039.2 | XP_014973525.1 | 471 | 77 | ||
| XM_015118040.2 | XP_014973526.1 | 480 | 77 | |||
| XM_015118041.2 | XP_014973527.1 | 392 | 77 | |||
| XM_015118042.2 | XP_014973528.1 | 432 | 77 | |||
| XM_015118043.2 | XP_014973529.1 | 1534 | 77 | |||
| XM_015118045.2 | XP_014973531.1 | 309 | 77 | |||
| XM_028835305.1 | XP_028691138.1 | 281 | 77 | |||
| XM_028835306.1 | XP_028691139.1 | 478 | 77 | |||
| XM_028835307.1 | XP_028691140.1 | 609 | 77 | |||
| GenBank mRNA | JU332641.1 | - | 298 | 77 | ||
| JU472154.1 | - | 381 | 77 | |||
| JV044211.1 | - | 367 | 77 | |||
| JV044212.1 | - | 367 | 77 | |||
| JV046008.1 | - | 367 | 77 | |||
Set Membership: |
|