Search
Search

ANKRD29| Gene Symbol: ANKRD29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121676.2 | XP_014977162.1 | 407 | 66 | ||
| XM_015121677.2 | XP_014977163.1 | 381 | 66 | |||
| XM_015121678.2 | XP_014977164.1 | 381 | 66 | |||
| XM_015121679.2 | XP_014977165.1 | 371 | 66 | |||
| XM_028837883.1 | XP_028693716.1 | 381 | 66 | |||
| XM_028837884.1 | XP_028693717.1 | 137 | 66 | |||
| GenBank mRNA | JU322046.1 | - | 239 | 66 | ||
Set Membership: |
|