Search
Search

DDR2| Gene Symbol: DDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114086.2 | XP_014969572.1 | 3160 | 102 | ||
| XM_015114088.2 | XP_014969574.1 | 3040 | 102 | |||
| XM_028826657.1 | XP_028682490.1 | 3050 | 102 | |||
| GenBank mRNA | JU327122.1 | - | 2287 | 102 | ||
| JV062179.1 | - | 2216 | 102 | |||
Set Membership: |
|