Search
Search

SMARCA2| Gene Symbol: SMARCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117810.2 | XP_014973296.1 | 2500 | 87 | ||
| XM_015117811.2 | XP_014973297.1 | 2652 | 87 | |||
| XM_015117813.2 | XP_014973299.1 | 2755 | 87 | |||
| XM_015117817.2 | XP_014973303.1 | 2266 | 87 | |||
| XM_015117818.2 | XP_014973304.1 | 2245 | 87 | |||
| XM_028835200.1 | XP_028691033.1 | 3488 | 87 | |||
| XM_028835201.1 | XP_028691034.1 | 2555 | 87 | |||
| XM_028835202.1 | XP_028691035.1 | 2433 | 87 | |||
| XM_028835203.1 | XP_028691036.1 | 2433 | 87 | |||
| GenBank mRNA | JU328367.1 | - | 1456 | 87 | ||
| JU328368.1 | - | 1456 | 87 | |||
| JV591476.1 | - | 1486 | 87 | |||
Set Membership: |
|