Search
Search

HERC1| Gene Symbol: HERC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142428.2 | XP_014997914.2 | 12957 | 91 | ||
| XM_015142434.2 | XP_014997920.2 | 12930 | 91 | |||
| XM_015142436.2 | XP_014997922.2 | 12879 | 91 | |||
| XM_028851017.1 | XP_028706850.1 | 13023 | 91 | |||
| XM_028851018.1 | XP_028706851.1 | 13020 | 91 | |||
| XM_028851019.1 | XP_028706852.1 | 13002 | 91 | |||
| XM_028851020.1 | XP_028706853.1 | 12999 | 91 | |||
| XM_028851022.1 | XP_028706855.1 | 12996 | 91 | |||
| XM_028851023.1 | XP_028706856.1 | 12972 | 91 | |||
| GenBank mRNA | JU322252.1 | - | 12702 | 91 | ||
| JU336957.1 | - | 12681 | 91 | |||
| JU477690.1 | - | 12739 | 91 | |||
| JU477691.1 | - | 12718 | 91 | |||
| JV485993.1 | - | 10102 | 91 | |||
Set Membership: |
|