Search
Search

XPO1| Gene Symbol: XPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266844.1 | NP_001253773.1 | 3448 | 145 | ||
| XM_028831640.1 | XP_028687473.1 | 3408 | 145 | |||
| XM_028831641.1 | XP_028687474.1 | 3916 | 145 | |||
| XM_028831642.1 | XP_028687475.1 | 2644 | 145 | |||
| GenBank mRNA | JU319856.1 | - | 3024 | 145 | ||
| JU472665.1 | - | 3485 | 145 | |||
| JV044253.1 | - | 3448 | 145 | |||
| JV637829.1 | - | 3482 | 145 | |||
Set Membership: |
|