Search
Search

CDK19| Gene Symbol: CDK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137573.2 | XP_014993059.1 | 1287 | 92 | ||
| XM_015137574.2 | XP_014993060.1 | 1155 | 92 | |||
| XM_015137575.2 | XP_014993061.1 | 1119 | 92 | |||
| XM_015137577.2 | XP_014993063.1 | 980 | 92 | |||
| XM_015137578.2 | XP_014993064.1 | 946 | 92 | |||
| XM_028847026.1 | XP_028702859.1 | 1125 | 92 | |||
| XM_028847027.1 | XP_028702860.1 | 1289 | 92 | |||
| XM_028847028.1 | XP_028702861.1 | 1295 | 92 | |||
| XM_028847029.1 | XP_028702862.1 | 986 | 92 | |||
| XM_028847030.1 | XP_028702863.1 | 1097 | 92 | |||
| XM_028847031.1 | XP_028702864.1 | 685 | 92 | |||
| GenBank mRNA | JU333540.1 | - | 914 | 92 | ||
| JU333541.1 | - | 914 | 92 | |||
| JU474806.1 | - | 927 | 92 | |||
| JU474807.1 | - | 927 | 92 | |||
| JV646041.1 | - | 937 | 92 | |||
Set Membership: |
|