Search
Search

MNAT1| Gene Symbol: MNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261796.1 | NP_001248725.1 | 173 | 119 | ||
| XM_028850307.1 | XP_028706140.1 | 227 | 119 | |||
| XM_028850308.1 | XP_028706141.1 | 227 | 119 | |||
| XM_028850309.1 | XP_028706142.1 | 227 | 119 | |||
| GenBank mRNA | JU320636.1 | - | 208 | 119 | ||
| JU473378.1 | - | 173 | 119 | |||
Set Membership: |
|