Search
Search

USP40| Gene Symbol: USP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111376.2 | XP_014966862.1 | 3603 | 89 | ||
| XM_015111377.2 | XP_014966863.1 | 3588 | 89 | |||
| XM_015111378.2 | XP_014966864.1 | 3603 | 89 | |||
| XM_015111380.2 | XP_014966866.1 | 3170 | 89 | |||
| XM_028831251.1 | XP_028687084.1 | 3739 | 89 | |||
| GenBank mRNA | JU489167.1 | - | 3439 | 89 | ||
Set Membership: |
|