Search
Search

KAT6B| Gene Symbol: KAT6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002805674.3 | XP_002805720.3 | 3676 | 71 | ||
| XM_015147237.2 | XP_015002723.2 | 4230 | 71 | |||
| XM_015147238.2 | XP_015002724.2 | 3819 | 71 | |||
| XM_015147239.2 | XP_015002725.2 | 3768 | 71 | |||
| XM_015147240.2 | XP_015002726.2 | 3966 | 71 | |||
| XM_015147242.2 | XP_015002728.2 | 3218 | 71 | |||
| GenBank mRNA | JU341190.1 | - | 104 | 71 | ||
| JU483576.1 | - | 1844 | 71 | |||
Set Membership: |
|