Search
Search

EIF2A| Gene Symbol: EIF2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131799.2 | XP_014987285.1 | 675 | 58 | ||
| XM_015131800.2 | XP_014987286.1 | 555 | 58 | |||
| XM_028843776.1 | XP_028699609.1 | 498 | 58 | |||
| GenBank mRNA | JU321089.1 | - | 574 | 58 | ||
| JV048406.1 | - | 572 | 58 | |||
| JV724787.1 | - | 572 | 58 | |||
Set Membership: |
|