Search
Search

AMPH| Gene Symbol: AMPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100038.4 | XP_001100038.1 | 363 | 92 | ||
| XM_001100126.4 | XP_001100126.2 | 363 | 92 | |||
| GenBank mRNA | JU334026.1 | - | 212 | 92 | ||
| JV541369.1 | - | 219 | 92 | |||
| JV565101.1 | - | 211 | 92 | |||
| JV667379.1 | - | 206 | 92 | |||
Set Membership: |
|