Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh01048244_mH
See other ABI2 GE Assays ›
Gene Symbol:
ABI2
Gene Name:
abl interactor 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 90715790 - 90831582 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh01048244_mH
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
4,430.00
Your Price
Online offer:
4430.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_001102942.4
XP_001102942.1
1642
118
XM_001103104.4
XP_001103104.2
1711
118
XM_001103265.4
XP_001103265.2
1624
118
XM_001103414.4
XP_001103414.2
1441
118
XM_001103497.4
XP_001103497.1
1528
118
XM_015110752.2
XP_014966238.1
1730
118
XM_015110753.2
XP_014966239.1
1726
118
XM_015110754.2
XP_014966240.1
1711
118
XM_015110755.2
XP_014966241.1
1642
118
XM_015110756.2
XP_014966242.1
1542
118
XM_015110757.2
XP_014966243.1
1582
118
XM_015110758.2
XP_014966244.1
1579
118
XM_015110759.2
XP_014966245.1
1564
118
XM_015110760.2
XP_014966246.1
1546
118
XM_015110761.2
XP_014966247.1
1528
118
XM_015110762.2
XP_014966248.1
1546
118
XM_015110763.2
XP_014966249.1
1525
118
XM_015110765.2
XP_014966251.1
1459
118
XM_015110766.2
XP_014966252.1
1477
118
XM_015110767.2
XP_014966253.1
1456
118
XM_015110769.2
XP_014966255.1
1441
118
XM_015110770.2
XP_014966256.1
1459
118
XM_028831006.1
XP_028686839.1
1726
118
XM_028831008.1
XP_028686841.1
1561
118
XM_028831009.1
XP_028686842.1
1579
118
XM_028831010.1
XP_028686843.1
1460
118
XM_028831011.1
XP_028686844.1
1492
118
XM_028831012.1
XP_028686845.1
1378
118
XM_028831013.1
XP_028686846.1
1312
118
GenBank mRNA
JU320691.1
-
1478
118
JU332266.1
-
1478
118
JU332267.1
-
1478
118
JU332268.1
-
1478
118
JU332269.1
-
1478
118
JU332270.1
-
1478
118
JU332271.1
-
1478
118
JU473890.1
-
1273
118
JV043999.1
-
1460
118
JV637513.1
-
1487
118
Back To Top
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Recommended Assay
Macaca mulatta
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit