Search
Search

XYLT1| Gene Symbol: XYLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125707.2 | XP_014981193.1 | 1446 | 90 | ||
| GenBank mRNA | JU326352.1 | - | 1062 | 90 | ||
| JV625506.1 | - | 1138 | 90 | |||
Set Membership: |
|