Search
Search

PHKB| Gene Symbol: PHKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261044.1 | NP_001247973.1 | 1180 | 132 | ||
| XM_015126047.2 | XP_014981533.1 | 1115 | 132 | |||
| XM_015126049.2 | XP_014981535.1 | 1087 | 132 | |||
| XM_015126050.2 | XP_014981536.1 | 1262 | 132 | |||
| XM_028840584.1 | XP_028696417.1 | 1097 | 132 | |||
| XM_028840585.1 | XP_028696418.1 | 1233 | 132 | |||
| XM_028840586.1 | XP_028696419.1 | 1392 | 132 | |||
| XM_028840587.1 | XP_028696420.1 | 1033 | 132 | |||
| XM_028840588.1 | XP_028696421.1 | 795 | 132 | |||
| GenBank mRNA | JU322067.1 | - | 1135 | 132 | ||
| JU322068.1 | - | 1135 | 132 | |||
| JU473668.1 | - | 1197 | 132 | |||
| JV044061.1 | - | 1197 | 132 | |||
| JV047374.1 | - | 1180 | 132 | |||
| JV047375.1 | - | 1085 | 132 | |||
Set Membership: |
|