Search
Search

COMTD1| Gene Symbol: COMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261688.1 | NP_001248617.1 | 520 | 150 | ||
| XM_028853377.1 | XP_028709210.1 | 597 | 150 | |||
| GenBank mRNA | JU332859.1 | - | 496 | 150 | ||
| JU474259.1 | - | 520 | 150 | |||
| JV605436.1 | - | 522 | 150 | |||
Set Membership: |
|