Search
Search

DOCK1| Gene Symbol: DOCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826998.1 | XP_028682831.1 | 299 | 69 | ||
| XM_028826999.1 | XP_028682832.1 | 500 | 69 | |||
| GenBank mRNA | JU320407.1 | - | 388 | 69 | ||
| JU320408.1 | - | 388 | 69 | |||
| JV636572.1 | - | 388 | 69 | |||
| JV704228.1 | - | 340 | 69 | |||
Set Membership: |
|