Search
Search

TCF7L1| Gene Symbol: TCF7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083116.4 | XP_001083116.3 | 1137 | 66 | ||
| XM_028832347.1 | XP_028688180.1 | 1099 | 66 | |||
| GenBank mRNA | JV662839.1 | - | 812 | 66 | ||
Set Membership: |
|