Search
Search

AXL| Gene Symbol: AXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839773.1 | XP_028695606.1 | 2425 | 79 | ||
| XM_028839774.1 | XP_028695607.1 | 2374 | 79 | |||
| XM_028839775.1 | XP_028695608.1 | 2098 | 79 | |||
| GenBank mRNA | JV043906.1 | - | 2127 | 79 | ||
| JV043907.1 | - | 2100 | 79 | |||
| JV635405.1 | - | 1983 | 79 | |||
Set Membership: |
|